Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335258

Sigma-Aldrich

trans-Nerolidol

≥98% (HPLC)

Synonym(s):

(E)-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, trans-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
222.37
Beilstein/REAXYS Number:
5731231
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

≥98% (HPLC)

refractive index

n20/D 1.479 (lit.)

bp

145-146 °C/12 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C/CC\C(C)=C\CCC(C)(O)C=C

InChI

1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11+

InChI key

FQTLCLSUCSAZDY-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service