Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

334774

Sigma-Aldrich

N-Methylhomoveratrylamine

97%

Synonym(s):

3,4-Dimethoxy-N-methylphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2NHCH3
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

drug control

USDEA Schedule I

refractive index

n20/D 1.533 (lit.)

bp

160-161 °C/13 mmHg (lit.)

density

1.059 g/mL at 25 °C (lit.)

SMILES string

CNCCc1ccc(OC)c(OC)c1

InChI

1S/C11H17NO2/c1-12-7-6-9-4-5-10(13-2)11(8-9)14-3/h4-5,8,12H,6-7H2,1-3H3

InChI key

HNJWKRMESUMDQE-UHFFFAOYSA-N

Application

N-Methylhomoveratrylamine was used in the preparation of:
  • 5,6-dimethoxy-1-oxo-isoindoline derivatives[1]
  • N-{4-[2-hydroxy-3-(3,4-dimethoxy-N-methylphenylethylamino)propoxy]phenyl}-5,6-dimethoxy-1-oxo-isoindoline[1]
  • cis[3-[2-[2-(3,4-dimethoxyphenyl)ethyl]methylamino]cyclohexyl]diphenylacetonitrile[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dharam P Jindal et al.
Bioorganic chemistry, 33(4), 310-324 (2005-06-25)
Phenoxypropanolamines with 1-oxo-isoindoline and 5,6-dimethoxy-1-oxo-isoindoline groups at the para position were synthesized. beta1, beta2-Adrenergic receptor binding affinities for the synthesized compounds were tested and compared with propranolol and atenolol. It was found that the incorporation of para-amidic functionality within the
Silvia Dei et al.
Bioorganic & medicinal chemistry, 13(4), 985-998 (2005-01-27)
A series of compounds with a diphenylmethyl cyclohexyl skeleton, loosely related to verapamil, has been synthesized and tested as MDR modulators on anthracycline-resistant erythroleukemia K 562 cells. Their residual cardiovascular action (negative inotropic and chronotropic activity as well as vasorelaxant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service