Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

334723

Sigma-Aldrich

4,5-Dichlorophthalic anhydride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H2Cl2O3
CAS Number:
Molecular Weight:
217.01
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

98%

mp

185-187 °C (lit.)

SMILES string

Clc1cc2C(=O)OC(=O)c2cc1Cl

InChI

1S/C8H2Cl2O3/c9-5-1-3-4(2-6(5)10)8(12)13-7(3)11/h1-2H

InChI key

ULSOWUBMELTORB-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gayan S Jayawickrama et al.
PloS one, 13(4), e0196404-e0196404 (2018-04-25)
The mammalian kynurenine aminotransferase (KAT) enzymes are a family of related isoforms that are pyridoxal 5'-phosphate-dependent, responsible for the irreversible transamination of kynurenine to kynurenic acid. Kynurenic acid is implicated in human diseases such as schizophrenia where it is found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service