About This Item
Recommended Products
vapor pressure
1 mmHg ( 970 °C)
assay
98%
composition
Degree of hydration, 3-4
SMILES string
O.F[Zn]F
InChI
1S/2FH.H2O.Zn/h2*1H;1H2;/q;;;+2/p-2
InChI key
OZHOVPJDYIFXTJ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service