Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

332917

Sigma-Aldrich

Aluminum fluoride

reagent grade

Synonym(s):

Aluminum trifluoride, Trifluoroaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlF3
CAS Number:
Molecular Weight:
83.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

reagent grade

form

powder

mp

1290 °C (lit.)
1291 °C (lit.)

density

3.1 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

F[Al](F)F

InChI

1S/Al.3FH/h;3*1H/q+3;;;/p-3

Inchi Key

KLZUFWVZNOTSEM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia C Burrows et al.
Nucleic acids research, 37(15), 5138-5150 (2009-06-26)
Bacterial RNA polymerase (RNAP) containing the major variant sigma(54) factor forms open promoter complexes in a reaction in which specialized activator proteins hydrolyse ATP. Here we probe binding interactions between sigma(54)-RNAP (Esigma(54)) and the ATPases associated with various cellular activities
Lumei Liu et al.
Dalton transactions (Cambridge, England : 2003), 39(7), 1722-1725 (2010-05-08)
AlF(bdc), bdc = 1,4-benzenedicarboxylate, a new member of the family of MX(bdc) compounds has been synthesized under solvothermal conditions. It is formed without H(2)bdc in the channels and substantial absorption capacity is observed in its as-synthesized form. AlF(bdc) does not
Dinesh Shetty et al.
Chemical communications (Cambridge, England), 47(34), 9732-9734 (2011-07-22)
A single crystal structure of an aluminium-fluoride complex of a model compound (NODA-benzyl) was studied to understand the co-ordination chemistry. Series of ligands with an extra carboxylic acid linker for biomolecule conjugation were studied for improved (18)F-labeling applications.
Xiaoyan Jin et al.
Environmental science & technology, 45(1), 288-293 (2010-12-08)
Density functional theory (DFT) calculation is carried out to investigate the structures, (19)F and (27)Al NMR chemical shifts of aqueous Al-F complexes and their water-exchange reactions. The following investigations are performed in this paper: (1) the microscopic properties of typical
Peter Laverman et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 33(2), 427-434 (2011-10-20)
We recently reported a facile method based on the chelation of [(18)F]aluminum fluoride (Al(18)F) by NOTA (1,4,7-triazacyclononane-1,4,7-triacetic acid). Here, we present a further optimization of the (18)F labeling of NOTA-octreotide (IMP466). Octreotide was conjugated with the NOTA chelate and was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service