Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

331953

Sigma-Aldrich

Tetrabutylammonium fluoride on alumina

extent of labeling: 15 wt. % loading

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(F)
CAS Number:
Molecular Weight:
261.46
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

extent of labeling

15 wt. % loading

SMILES string

[F-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.FH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

FPGGTKZVZWFYPV-UHFFFAOYSA-M

Application

  • Catalyst for etherification of alcohols and phenols with alkyl halides
  • Catalyst for benzylation reactions

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

supp_hazards

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Zhong et al.
Carbohydrate polymers, 94(1), 38-45 (2013-04-03)
In this article, a novel and high efficient solvent, tetra-n-Butylammonium Hydroxide (TBAH), was used for dissolution and isolation of straw cellulose from wheat straw. The composition analysis with gas chromatography (GC) and the spectroscopic characterization analysis conducted by X-Ray diffraction
Oliver Bixner et al.
The Journal of chemical physics, 136(20), 204503-204503 (2012-06-07)
The interaction of exciton and charge transfer (CT) states plays a central role in photo-induced CT processes in chemistry, biology, and physics. In this work, we use a combination of two-dimensional electronic spectroscopy (2D-ES), pump-probe measurements, and quantum chemistry to
Mei Shen et al.
Journal of the American Chemical Society, 134(24), 9856-9859 (2012-06-05)
Here we report on the unprecedentedly high resolution imaging of ion transport through single nanopores by scanning electrochemical microscopy (SECM). The quantitative SECM image of single nanopores allows for the determination of their structural properties, including their density, shape, and
R Bryan Sears et al.
Journal of inorganic biochemistry, 121, 77-87 (2013-01-29)
The complex cis-[Ru(phpy)(phen)(CH3CN)2](+) (phpy=2-phenylpyridine, phen=1,10-phenanthroline) was investigated as a potential photodynamic therapy (PDT) agent. This complex presents desirable photochemical characteristics including a low energy absorption tail extending into the PDT window (600-850nm) and photoinduced exchange of the CH3CN ligands, generating
Erbo Shi et al.
Organic letters, 14(13), 3384-3387 (2012-06-27)
A metal-free C-H oxidation for the construction of allylic esters has been developed. The use of a commercially available and inexpensive catalyst and oxidant, and readily available starting materials, coupled with the operational simplicity of the reaction, renders the methodology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service