330698
Bis(trimethylsilyl)cyclopentadiene,mixture of isomers
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H22Si2
CAS Number:
Molecular Weight:
210.46
UNSPSC Code:
12352103
assay
98%
refractive index
n20/D 1.477 (lit.)
density
0.851 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)C1C=CC(=C1)[Si](C)(C)C
Other Notes
Isomer ratio varies with temperature
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service