Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330450

Sigma-Aldrich

p-Toluidine hydrochloride

98%

Synonym(s):

4-Methylaniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2 · HCl
CAS Number:
Molecular Weight:
143.61
Beilstein/REAXYS Number:
3557540
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

243-245 °C (lit.)

SMILES string

Cl[H].Cc1ccc(N)cc1

InChI

1S/C7H9N.ClH/c1-6-2-4-7(8)5-3-6;/h2-5H,8H2,1H3;1H

InChI key

JQKBUTDZZRGQDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service