Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328499

Sigma-Aldrich

N-Ethyl-4-nitroaniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NHC2H5
CAS Number:
Molecular Weight:
166.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

mp

96-98 °C (lit.)

SMILES string

CCNc1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H10N2O2/c1-2-9-7-3-5-8(6-4-7)10(11)12/h3-6,9H,2H2,1H3

InChI key

XBNNLAWQCMDISJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonlinear optical properties and molecular orientation of the mixed crystals of p-nitroaniline and N-ethyl-4-nitroaniline was studied.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Second-Order Nonlinearity of Mixtures Including p-Nitroaniline Derivatives.
Kato M, et al.
The Journal of Physical Chemistry B, 101(44), 8856-8859 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service