Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328391

Sigma-Aldrich

3-Fluoro-3-deoxy-D-glucose

99%

Synonym(s):

3-Deoxy-3-fluoro-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11FO5
CAS Number:
Molecular Weight:
182.15
Beilstein/REAXYS Number:
1865171
MDL number:
UNSPSC Code:
12352103

assay

99%

optical activity

[α]25/D +61°, c = 0.7 in H2O

mp

115-117 °C (lit.)

SMILES string

OC[C@H]1OC(O)[C@H](O)[C@@H](F)[C@@H]1O

InChI

1S/C6H11FO5/c7-3-4(9)2(1-8)12-6(11)5(3)10/h2-6,8-11H,1H2/t2-,3+,4-,5-,6?/m1/s1

InChI key

BUMRBAMACDBPKO-CBPJZXOFSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service