326747
2,2,2-Trifluoroethanol
NMR grade, ≥99.5%
Synonym(s):
Trifluoroethyl alcohol
About This Item
Recommended Products
vapor density
3.5 (vs air)
vapor pressure
70 mmHg ( 25 °C)
assay
≥99.5%
autoignition temp.
~896 °F
expl. lim.
42 %
refractive index
n20/D 1.3 (lit.)
bp
77-80 °C (lit.)
mp
−44 °C (lit.)
density
1.373 g/mL at 25 °C (lit.)
SMILES string
OCC(F)(F)F
InChI
1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2
InChI key
RHQDFWAXVIIEBN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation
target_organs
Blood
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
86.0 °F - closed cup
flash_point_c
30 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service