Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

326305

Sigma-Aldrich

Trichlorofluoromethane

NMR grade, ≥99.5%

Synonym(s):

CFC-11, Fluorotrichloromethane, Freon® 11

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CF
CAS Number:
Molecular Weight:
137.37
Beilstein/REAXYS Number:
1732469
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

5.04 (vs air)

vapor pressure

12.85 psi ( 20 °C)
39.17 psi ( 55 °C)

assay

≥99.5%

refractive index

n20/D 1.3821 (lit.)

bp

23.7 °C (lit.)

mp

−111-−110 °C (lit.)

density

1.494 g/mL at 25 °C (lit.)

SMILES string

FC(Cl)(Cl)Cl

InChI

1S/CCl3F/c2-1(3,4)5

InChI key

CYRMSUTZVYGINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in ampules

Legal Information

Freon is a registered trademark of E. I. du Pont de Nemours and Company

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Ozone 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service