326119
Hafnium(IV) oxide
−325 mesh, 99.9% (Purity excludes 0.5-1% zirconium)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
HfO2
CAS Number:
Molecular Weight:
210.49
MDL number:
assay
99.9% (Purity excludes 0.5-1% zirconium)
particle size
−325 mesh
density
9.68 g/mL at 25 °C (lit.)
SMILES string
O=[Hf]=O
InChI
1S/Hf.2O
InChI key
CJNBYAVZURUTKZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for spectrographic applications
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service