325457
Copper(II) oxide
≥99.99%
Synonym(s):
Cupric oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352300
Recommended Products
assay
≥99.99%
SMILES string
[Cu]=O
InChI
1S/Cu.O
InChI key
QPLDLSVMHZLSFG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
Product No.
Description
Pricing
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sumanta Kumar Meher et al.
Nanoscale, 5(5), 2089-2099 (2013-02-06)
In the quest to enhance the selectivity and sensitivity of novel structured metal oxides for electrochemical non-enzymatic sensing of glucose, we report here a green synthesis of unique sandwich-structured CuO on a large scale under microwave mediated homogeneous precipitation conditions.
Chengyi Hou et al.
Nanoscale, 5(3), 1227-1232 (2013-01-11)
We report a Cu(2)O nanocrystal-reduced graphene oxide hybrid that is dispersible in water and has anticancer activity under both visible and near-infrared light irradiation. In contrast to the highly efficient killing of both normal and cancer cells initiated by the
Nicola Scotti et al.
Chemical communications (Cambridge, England), 49(19), 1957-1959 (2013-01-30)
The epoxidation of alkenes was carried out over a CuO/Al(2)O(3) catalyst using cumene as an oxygen carrier, through a one-pot reaction, giving high conversion and selectivity with different substrates. Trans-β-methylstyrene gave the corresponding epoxide in 95% yield after 3 h.
Yifu Yu et al.
Nanoscale, 5(3), 904-909 (2012-12-21)
A simple and feasible contact mode called gravitational contact mode (GCM) was developed for the first time to imitate the practical state between soot and catalyst. By simulating rainwater adsorption on a lawn in nature, we synthesized a lawn-like CuO
Yunxin Liu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4319-4326 (2013-03-01)
How to extend ultraviolet photocatalysts to the visible-light region is a key challenge for solar-driven photocatalysis. Herein, we show that ultraviolet ZnO photocatalysts can present high visible-light photocatalytic activity when combined with CuO quantum dots (QDs; <3 nm). Theoretical analysis demonstrates
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service