323608
3′-Azido-3′-deoxythymidine
98%
Synonym(s):
AZT, Azidothymidine, ZDV, Zidovudine
About This Item
Recommended Products
assay
98%
optical activity
[α]20/D +47°, c = 0.5 in H2O
mp
113-115 °C (lit.)
SMILES string
CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
InChI
1S/C10H13N5O4/c1-5-3-15(10(18)12-9(5)17)8-2-6(13-14-11)7(4-16)19-8/h3,6-8,16H,2,4H2,1H3,(H,12,17,18)/t6-,7+,8+/m0/s1
InChI key
HBOMLICNUCNMMY-XLPZGREQSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service