323020
Ethyl 3-(1-azetidinyl)propionate
97%
Synonym(s):
1-(2-Carbethoxyethyl)azetidine, 1-(2-Ethoxycarbonylethyl)azetidine, Ethyl azetidine-1-propionate
About This Item
assay
97%
refractive index
n20/D 1.441 (lit.)
bp
86-87 °C/12 mmHg (lit.)
density
0.965 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCOC(=O)CCN1CCC1
InChI
1S/C8H15NO2/c1-2-11-8(10)4-7-9-5-3-6-9/h2-7H2,1H3
InChI key
JHEBCRMYKMBVBX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
176.0 °F - closed cup
flash_point_c
80 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service