Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

322466

Sigma-Aldrich

Sodium pyrophosphate tetrabasic

98%

Synonym(s):

Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7
CAS Number:
Molecular Weight:
265.90
EC Number:
MDL number:
UNSPSC Code:
12352302

assay

98%

density

2.53 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2/c;;;;1-8(2,3)7-9(4,5)6/h;;;;(H2,1,2,3)(H2,4,5,6)/q4*+1;/p-4

InChI key

FQENQNTWSFEDLI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 19(7), 1855-1857 (2009-03-10)
An inhibition study of the cytosolic carbonic anhydrase (CA, EC 4.2.1.1) isoforms I, II, III, VII and XIII with anions such as stannate(IV), selenate(VI), tellurate(VI), perosmate(VIII), persulfate, pyrophosphate(V), pyrovanadate(V), tetraborate, persulfate, perrhenate(VII), perrutenate(VII), selenocyanate, iminodisulfonate, fluorosulfate and trithiocarbonate is reported.
Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 20(5), 1548-1550 (2010-02-09)
An inhibition study of the transmembrane carbonic anhydrase (CA, EC 4.2.1.1) isoforms IX, XII (tumor-associated), and XIV with anions such as stannate(IV), selenate(VI), tellurate(VI), perosmate(VIII), persulfate, pyrophosphate(V), pyrovanadate(V), tetraborate, persulfate, perrhenate(VII), perrutenate(VII), selenocyanate, iminodisulfonate, fluorosulfate, and trithiocarbonate is reported. Selenate
W Charles O'Neill et al.
Kidney international, 79(5), 512-517 (2010-12-03)
Pyrophosphate, which may be deficient in advanced renal failure, is a potent inhibitor of vascular calcification. To explore its use as a potential therapeutic, we injected exogenous pyrophosphate subcutaneously or intraperitoneally in normal rats and found that their plasma pyrophosphate
Jesús Torres et al.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft, 200, 126-133 (2015-05-02)
The aim of this study was to test bone tissue response to monetite granules in comparison with intramembranous autologous bone graft in a rabbit calvaria critical size defect model. Novel monetite granules were synthesized by thermal conversion of set brushite
Hironori Suzuki et al.
Nature structural & molecular biology, 22(7), 572-580 (2015-06-02)
Atg101 is an essential component of the autophagy-initiating ULK complex in higher eukaryotes, but it is absent from the functionally equivalent Atg1 complex in budding yeast. Here, we report the crystal structure of the fission yeast Atg101-Atg13 complex. Atg101 has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service