Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

321265

Sigma-Aldrich

Carbon monoxide

10 ppm in nitrogen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO
CAS Number:
Molecular Weight:
28.01
Beilstein/REAXYS Number:
3587264
MDL number:
Pricing and availability is not currently available.

autoignition temp.

1128 °F

expl. lim.

74 %

concentration

10 ppm in nitrogen

bp

−191.5 °C (lit.)

mp

−205 °C (lit.)

SMILES string

[C-]#[O+]

InChI

1S/CO/c1-2

InChI key

UGFAIRIUMAVXCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For calibrating analytical instruments.

Packaging

Packaged in lightweight no-return cylinder

Recommended products

Control valve Z166995 or regulator Z167002 is recommended.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service