319597
Thymol Blue solution
0.04 wt. % in H2O
Synonym(s):
Thymolsulfonphthalein sodium salt
About This Item
Recommended Products
concentration
0.04 wt. % in H2O
pH
1.2-2.8, red to yellow
density
0.979 g/mL at 25 °C
λmax
590 nm
SMILES string
[Na+].CC(C)c1cc(c(C)cc1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c([O-])cc4C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service