Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

319562

Supelco

Sodium sulfate solution

in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
Beilstein/REAXYS Number:
3648446
MDL number:
UNSPSC Code:
12000000

concentration

in H2O

density

1 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Temperini et al.
Bioorganic & medicinal chemistry letters, 17(10), 2795-2801 (2007-03-10)
N-Hydroxysulfamide is a 2000-fold more potent inhibitor of the zinc enzyme carbonic anhydrase (CA, EC 4.2.1.1) as compared to sulfamide. It also inhibits other physiologically relevant isoforms, such as the tumor-associated CA IX and XII (K(I)s in the range of
Deniz Ekinci et al.
Bioorganic & medicinal chemistry, 19(2), 744-748 (2011-01-08)
Carbonic anhydrase (CA; EC 4.2.1.1) was purified from the gill of the teleost fish Dicentrarchus labrax (European seabass). The purification procedure consisted of a single step affinity chromatography on Sepharose 4B-tyrosine-sulfanilamide. The enzyme was purified 84.9-fold with a yield of
Peter Burghout et al.
Bioorganic & medicinal chemistry, 19(1), 243-248 (2010-12-18)
The Gram-positive bacterium Streptococcus pneumoniae is a human respiratory tract pathogen that contributes significantly to global mortality and morbidity. It was recently shown that this bacterial pathogen depends on a conserved β-carbonic anhydrase (CA, EC 4.2.1.1) for in vitro growth
Daniela Vullo et al.
Bioorganic & medicinal chemistry letters, 21(12), 3591-3595 (2011-05-17)
Two new β-carbonic anhydrases (CAs, EC 4.2.1.1) from the bacterial pathogen Salmonella enterica serovar Typhimurium, stCA 1 and stCA 2, were characterized kinetically. The two enzymes possess appreciable activity as catalysts for the hydration of CO(2) to bicarbonate, with k(cat)
Anna Ohradanova et al.
Bioorganic & medicinal chemistry letters, 22(3), 1314-1316 (2012-01-10)
An α-carbonic anhydrase (CA, EC 4.2.1.1) isolated from the living fossil sponge Astrosclera willeyana, Astrosclerin, was investigated for its inhibition profile with simple inorganic anions, complex anions and other small molecules known to interact with these zinc enzymes. Astrosclerin is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service