Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

317403

Sigma-Aldrich

4-Chloro-1-naphthol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC10H6OH
CAS Number:
Molecular Weight:
178.61
Beilstein/REAXYS Number:
509750
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

99%

mp

118-121 °C (lit.)

SMILES string

Oc1ccc(Cl)c2ccccc12

InChI

1S/C10H7ClO/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6,12H

InChI key

LVSPDZAGCBEQAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Peroxidase stain.[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Houen et al.
Analytical biochemistry, 204(2), 296-299 (1992-08-01)
A specific peroxidase-coupled activity staining method for diamine oxidase (DAO) was developed. Diaminobenzidine was found to inhibit DAO and to give rise to unspecific staining. Among several other reagents 4-Cl-1-naphthol was found to be most suitable. Using specific activity staining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service