Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

316504

Millipore

Pentane

PRA grade, 98.0% n-pentane basis, >99.5% C5 isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein/REAXYS Number:
969132
EC Number:
MDL number:
UNSPSC Code:
12190000

grade

PRA grade

vapor density

2.48 (vs air)

vapor pressure

26.98 psi ( 55 °C)
8.4 psi ( 20 °C)

assay

98.0% n-pentane basis

autoignition temp.

500 °F

expl. lim.

~8.3 %

impurities

≤0.01% water

evapn. residue

<0.0001%

halogenated residue (GC/ECD)

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for pesticide residue analysis

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-40.0 °F

flash_point_c

-40 °C

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service