311049
Isopentylbenzene
97%
Synonym(s):
(3-Methyl-1-butyl)benzene, Isoamylbenzene
About This Item
assay
97%
bp
197 °C (lit.)
density
0.854 g/mL at 20 °C (lit.)
SMILES string
CC(C)CCc1ccccc1
InChI
1S/C11H16/c1-10(2)8-9-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3
InChI key
XNXIYYFOYIUJIW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
149.0 °F - closed cup
flash_point_c
65 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service