Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

309990

Sigma-Aldrich

Nickel(II) octanoate hydrate

98%

Synonym(s):

Octanoic acid nickel(II) salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)6CO2]2Ni · xH2O
CAS Number:
Molecular Weight:
345.10 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

powder

reaction suitability

core: nickel

SMILES string

O.CCCCCCCC(=O)O[Ni]OC(=O)CCCCCCC

InChI

1S/2C8H16O2.Ni.H2O/c2*1-2-3-4-5-6-7-8(9)10;;/h2*2-7H2,1H3,(H,9,10);;1H2/q;;+2;/p-2

InChI key

GOLWVMMVTHBFHR-UHFFFAOYSA-L

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service