Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

309710

Sigma-Aldrich

(R)-(+)-1,2,4-Butanetriol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
106.12
Beilstein/REAXYS Number:
4652491
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

viscous liquid

optical activity

[α]20/D +28±2, c = 1 in methanol

optical purity

ee: ≥96% (GLC)

bp

150 °C/0.04 mmHg (lit.)

density

1.19 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC[C@@H](O)CO

InChI

1S/C4H10O3/c5-2-1-4(7)3-6/h4-7H,1-3H2/t4-/m1/s1

InChI key

ARXKVVRQIIOZGF-SCSAIBSYSA-N

Application

(R)-(+)-1,2,4-Butanetriol can be used as a reactant to prepare:
  • Phenanthrene 9, 10 diacetal from phenanthrene 9, 10 quinone.[1]
  • (R)-1,2,4-Tris(benzoyloxy)butane by reacting with benzyl chloride in the presence of a base and DMAP.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct protection of 1,2 diols from α-diketones
Douglas NL, et al.
Synlett (1996)
Determination of the relative and absolute stereochemistry of fostriecin (CI-920)
Boger DL, et al.
The Journal of Organic Chemistry, 62(6), 1748-1753 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service