Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

308927

Sigma-Aldrich

4-Fluoroveratrole

97%

Synonym(s):

4-Fluoro-1,2-dimethoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3-1,2-(OCH3)2
CAS Number:
Molecular Weight:
156.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.51 (lit.)

density

1.171 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(F)cc1OC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tryfon Zarganes-Tzitzikas et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
Imaging techniques, such as positron emission tomography (PET), represent great progress in the clinical development of drugs and diagnostics. However, the efficient and timely synthesis of appropriately labeled compounds is a largely unsolved problem. Numerous small drug-like molecules with high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service