Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

308897

Sigma-Aldrich

Trifluoroacetic acid-d

99.5 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOD
CAS Number:
Molecular Weight:
115.03
Beilstein/REAXYS Number:
1768662
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.5 atom % D

mol wt

115.02 by atom % calculation

packaging

pk of 5 × 0.5 mL

impurities

≤0.500% water
water

refractive index

n20/D 1.3 (lit.)

bp

75 °C (lit.)

mp

-15 °C (lit.)

density

1.493 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)/i/hD

InChI key

DTQVDTLACAAQTR-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service