Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

308846

Sigma-Aldrich

Nitrobenzene-d5

99.5 atom % D

Synonym(s):

Pentadeuteronitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D5NO2
CAS Number:
Molecular Weight:
128.14
Beilstein/REAXYS Number:
1455693
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.5 atom % D

Quality Level

packaging

pk of 5 × 0.5 mL

refractive index

n20/D 1.5498 (lit.)

bp

88 °C/12 mmHg (lit.)

density

1.253 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])[N+]([O-])=O

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

LQNUZADURLCDLV-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1

target_organs

Blood

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

190.4 °F - closed cup

flash_point_c

88.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service