Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

308773

Sigma-Aldrich

Dichloromethane-d2

99.5 atom % D

Synonym(s):

Dideuteromethylenechloride, Methylene chloride-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2Cl2
CAS Number:
Molecular Weight:
86.94
Beilstein/REAXYS Number:
1733318
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
Product 308773 is not currently sold in your country. Contact Technical Service

vapor pressure

24.77 psi ( 55 °C)
7.06 psi ( 20 °C)

Quality Level

isotopic purity

99.5 atom % D

packaging

pk of 5 × 0.5 mL

refractive index

n20/D 1.422 (lit.)

bp

40 °C (lit.)

density

1.362 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(Cl)Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1D2

InChI key

YMWUJEATGCHHMB-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service