Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

306525

Sigma-Aldrich

Basacryl Red GL

Dye content 19 %

Synonym(s):

Basic Red 29

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClN4S
CAS Number:
Molecular Weight:
368.88
Colour Index Number:
11460
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 19%

λmax

511 nm

ε (extinction coefficient)

≥2800 at 275-285 nm in water at 0.009 g/L
≥6000 at 506-516 nm in water at 0.009 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cn1c(-c2ccccc2)c(\N=N\c3scc[n+]3C)c4ccccc14

InChI

1S/C19H17N4S.ClH/c1-22-12-13-24-19(22)21-20-17-15-10-6-7-11-16(15)23(2)18(17)14-8-4-3-5-9-14;/h3-13H,1-2H3;1H/q+1;/p-1

InChI key

WMAVHUWINYPPKT-UHFFFAOYSA-M

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Xia et al.
Cell, 176(6), 1379-1392 (2019-02-19)
Cell fate specification by lateral inhibition typically involves contact signaling through the Delta-Notch signaling pathway. However, whether this is the only signaling mode mediating lateral inhibition remains unclear. Here we show that in zebrafish oogenesis, a group of cells within

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service