Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

305898

Sigma-Aldrich

1-Chloro-2,3,5,6-tetrafluorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6HClF4
CAS Number:
Molecular Weight:
184.52
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

99%

SMILES string

Fc1cc(F)c(F)c(Cl)c1F

InChI

1S/C6HClF4/c7-4-5(10)2(8)1-3(9)6(4)11/h1H

InChI key

QLDHPPIOYFTGAJ-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

109.4 °F - closed cup

flash_point_c

43 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service