Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

305421

Sigma-Aldrich

3,4-Dihydro-6-methyl-2H-pyran-2-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.4677 (lit.)

bp

82-83 °C/15 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

CC1=CCCC(=O)O1

InChI

1S/C6H8O2/c1-5-3-2-4-6(7)8-5/h3H,2,4H2,1H3

InChI key

DOSOPEZOHKMQPJ-UHFFFAOYSA-N

General description

3,4-Dihydro-6-methyl-2H-pyran-2-one is an enol ether.

Application

3,4-Dihydro-6-methyl-2H-pyran-2-one was used as starting reagent in stereoselective synthesis of kasugamycin (antibiotic).

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The total synthesis of kasugamycin.
Y Suhara et al.
Journal of the American Chemical Society, 94(8), 6501-6507 (1972-09-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service