305294
(Trimethylsilyl)methanol
98%
Synonym(s):
(Hydroxymethyl)trimethylsilane
About This Item
assay
98%
refractive index
n20/D 1.419 (lit.)
bp
120-122 °C/754 mmHg (lit.)
density
0.826 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C[Si](C)(C)CO
InChI
1S/C4H12OSi/c1-6(2,3)4-5/h5H,4H2,1-3H3
InChI key
ZQKNBDOVPOZPLY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
132.8 °F - closed cup
flash_point_c
56 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service