Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

305243

Sigma-Aldrich

2-Mercapto-5-methylbenzimidazole

99%

Synonym(s):

5-Methyl-2-benzimidazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2S
CAS Number:
Molecular Weight:
164.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

mp

290-293 °C (lit.)

SMILES string

Cc1ccc2[nH]c(S)nc2c1

InChI

1S/C8H8N2S/c1-5-2-3-6-7(4-5)10-8(11)9-6/h2-4H,1H3,(H2,9,10,11)

InChI key

CWIYBOJLSWJGKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service