Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

301833

Sigma-Aldrich

4-Methyl-1,10-phenanthroline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
12352103

assay

99%

mp

143-145 °C (lit.)

SMILES string

Cc1ccnc2c1ccc3cccnc23

InChI

1S/C13H10N2/c1-9-6-8-15-13-11(9)5-4-10-3-2-7-14-12(10)13/h2-8H,1H3

InChI key

NAZZKEZTSOOCSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service