Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

301752

Sigma-Aldrich

2,4-Bis(5,6-diphenyl-1,2,4-triazin-3-yl)pyridine

98%

Synonym(s):

2,4-BDTP, 3,3′-(2,4-Pyridinediyl)bis(5,6-diphenyl-1,2,4-triazine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H23N7
CAS Number:
Molecular Weight:
541.60
Beilstein/REAXYS Number:
1194034
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

98%

SMILES string

c1ccc(cc1)-c2nnc(nc2-c3ccccc3)-c4ccnc(c4)-c5nnc(-c6ccccc6)c(n5)-c7ccccc7

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Artiss et al.
Clinical biochemistry, 14(6), 311-315 (1981-12-01)
A comparison of three sensitive ligands for iron is described. All show varying degrees of enhancement of values by copper, an element that is the primary interference encountered in serum assays for iron. Two of three tested reagents, 2,4-bis(5,6-diphenyl-1,2,4-triazin-3-yl) pyridine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service