Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

298646

Sigma-Aldrich

Octadecanophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO(CH2)16CH3
CAS Number:
Molecular Weight:
344.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

bp

425 °C (lit.)

mp

65-67 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)c1ccccc1

InChI

1S/C24H40O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19-22-24(25)23-20-17-16-18-21-23/h16-18,20-21H,2-15,19,22H2,1H3

InChI key

DBLXXVQTWJFJFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service