295256
1-Chloro-1,1-difluoroethane
98%
Synonym(s):
HCFC-142b
About This Item
Recommended Products
vapor density
3.49 (vs air)
vapor pressure
2196 mmHg ( 21 °C)
assay
98%
autoignition temp.
1169 °F
expl. lim.
18 %
bp
−10 °C (lit.)
mp
−131 °C (lit.)
SMILES string
CC(F)(F)Cl
InChI
1S/C2H3ClF2/c1-2(3,4)5/h1H3
InChI key
BHNZEZWIUMJCGF-UHFFFAOYSA-N
Application
Recommended products
hose barb
regulator
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Flam. Gas 1B - Ozone 1 - Press. Gas Liquefied gas
Storage Class
2A - Gases
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service