About This Item
Recommended Products
vapor density
4.05 (vs air)
Quality Level
vapor pressure
1128 mmHg ( 20 °C)
2815 mmHg ( 55 °C)
assay
99.9%
reaction suitability
reagent type: catalyst
core: boron
bp
12.5 °C (lit.)
mp
−107 °C (lit.)
SMILES string
ClB(Cl)Cl
InChI
1S/BCl3/c2-1(3)4
InChI key
FAQYAMRNWDIXMY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
recommended
signalword
Danger
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Press. Gas Compr. Gas - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
supp_hazards
Storage Class
2A - Gases
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service