Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

294845

Sigma-Aldrich

Bromomethane solution

2.0 M in diethyl ether

Synonym(s):

Methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Br
CAS Number:
Molecular Weight:
94.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1420 mmHg ( 20 °C)
3945 mmHg ( 55 °C)

Quality Level

concentration

2.0 M in diethyl ether

impurities

<0.005% water

density

0.83 g/mL at 25 °C

application(s)

agriculture

Storage temp.

2-8°C

SMILES string

CBr

InChI

1S/CH3Br/c1-2/h1H3

InChI key

GZUXJHMPEANEGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromomethane solution (methyl bromide) is the most effective soil fumigant for killing plant pests, its use is being phased out because its emissions from soil can lead to stratospheric ozone depletion[1]. The bimolecular nucleophilic substitution (SN2) reaction of methyl bromide and OH- in aqueous solution has been investigated using a multilayered-representation quantum mechanical and molecular mechanics methodology[2].

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Ozone 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-40.0 °F - closed cup

flash_point_c

-40 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richeng Xuan et al.
Environmental science & technology, 44(23), 9080-9085 (2010-11-10)
Despite generally being considered the most effective soil fumigant, methyl bromide (MeBr) use is being phased out because its emissions from soil can lead to stratospheric ozone depletion. However, a large amount is still currently used due to Critical Use
Khristy J Thompson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 120(1), 163-172 (2010-12-24)
Manganese (Mn) is an essential nutrient with potential neurotoxic effects. Mn deposited in the nose is apparently transported to the brain through anterograde axonal transport, bypassing the blood-brain barrier. However, the role of the olfactory epithelial cells in Mn transport
Guy J Hallman et al.
Journal of economic entomology, 104(1), 63-68 (2011-03-17)
Methyl bromide fumigation is widely used as a phytosanitary treatment. Mexican fruit fly, Anastrepha ludens (Loew) (Diptera: Tephritidae), is a quarantine pest of several fruit, including citrus (Citrus spp.), exported from Texas, Mexico, and Central America. Recently, live larvae have
Yulong Xu et al.
The Journal of chemical physics, 137(18), 184501-184501 (2012-11-21)
The bimolecular nucleophilic substitution (S(N)2) reaction of CH(3)Br and OH(-) in aqueous solution was investigated using a multilayered-representation quantum mechanical and molecular mechanics methodology. Reactant complex, transition state, and product complex are identified and characterized in aqueous solution. The potentials
Kathryn Hughes Barry et al.
Cancer causes & control : CCC, 23(6), 807-818 (2012-04-25)
Methyl bromide is a genotoxic soil fumigant with high acute toxicity, but unknown human carcinogenicity. Although many countries have reduced methyl bromide use because of its ozone depleting properties, some uses remain in the United States and other countries, warranting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service