29462
Dichloro-cyclohexyl-methylsilane
≥97.0% (GC)
Synonym(s):
Cyclohexyl-dichloro-methylsilane, Cyclohexyl-methyl-dichlorosilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H14Cl2Si
CAS Number:
Molecular Weight:
197.18
Beilstein/REAXYS Number:
2935173
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
≥97.0% (GC)
refractive index
n20/D 1.471
bp
83 °C/15 mmHg (lit.)
density
1.094 g/mL at 20 °C (lit.)
SMILES string
C[Si](Cl)(Cl)C1CCCCC1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service