293342
Octanoic hydrazide
technical grade, 80%
Synonym(s):
Caprylic hydrazide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)6CONHNH2
CAS Number:
Molecular Weight:
158.24
Beilstein/REAXYS Number:
1757322
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical grade
assay
80%
mp
87-89 °C (lit.)
SMILES string
CCCCCCCC(=O)NN
InChI
1S/C8H18N2O/c1-2-3-4-5-6-7-8(11)10-9/h2-7,9H2,1H3,(H,10,11)
InChI key
VMUZVGRNTPFTKE-UHFFFAOYSA-N
Application
Octanoic hydrazide (hydrazide of octanoic acid) has been used in the preparation of nitroamino- containing triazoles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Einarsson et al.
Journal of virological methods, 3(4), 213-228 (1981-11-01)
Deliberately added hepatitis B surface antigen (HBsAg) and hepatitis B virus (HBV) could be removed by hydrophobic interaction chromatography from a concentrate of coagulation factors II, VII, IX and X. Chromatography in high concentrations of salt, preferably on octanoic acid
New representatives of nitroamine-containing 1, 2, 4-triazoles: Synthesis and structure.
Efimova TP, et al.
Russ. J. Gen. Chem., 83(9), 1779-1786 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service