Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

292907

Sigma-Aldrich

Sulfathiazole

98%

Synonym(s):

2-Sulfanilamidothiazole, 4-Amino-N-(2-thiazolyl)benzenesulfonamide, N1-(2-Thiazolyl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O2S2
CAS Number:
Molecular Weight:
255.32
Beilstein/REAXYS Number:
226178
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

200-202 °C (lit.)

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2nccs2

InChI

1S/C9H9N3O2S2/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9/h1-6H,10H2,(H,11,12)

InChI key

JNMRHUJNCSQMMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Krátký et al.
European journal of medicinal chemistry, 50, 433-440 (2012-03-01)
A series of novel sulfonamides containing 5-chloro-2-hydroxybenzaldehyde or 5-chloro-2-hydroxybenzoic acid scaffolds were designed, synthesized and characterized by IR, (1)H NMR and (13)C NMR. All ten target synthesized derivatives and starting sulfonamides were evaluated in vitro for the activity against Gram-positive and
Tamas Hatfaludi et al.
Antimicrobial agents and chemotherapy, 52(11), 4166-4171 (2008-08-30)
Two TolC homologs, PM0527 and PM1980, were identified for Pasteurella multocida. A pm0527 mutant displayed increased susceptibility to a range of chemicals, including rifampin (512-fold) and acridine orange (128-fold). A pm1980 mutant showed increased susceptibility to rifampin, ceftazidime, and vancomycin.
Tony K L Kiang et al.
Pharmacology & therapeutics, 106(1), 97-132 (2005-03-23)
UDP-glucuronosyltransferase (UGT) enzymes catalyze the conjugation of various endogenous substances (e.g., bilirubin) and exogenous compounds (e.g., drugs). The human UGT superfamily is comprised of 2 families (UGT1 and UGT2) and 3 subfamilies (UGT1A, UGT2A, and UGT2B). Many of the individual
Minjun Chen et al.
Drug discovery today, 16(15-16), 697-703 (2011-06-01)
Drug-induced liver injury (DILI) is a leading cause of drugs failing during clinical trials and being withdrawn from the market. Comparative analysis of drugs based on their DILI potential is an effective approach to discover key DILI mechanisms and risk
Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service