Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

292885

Sigma-Aldrich

Guanidine thiocyanate

97%

Synonym(s):

Guanidinium rhodanide, Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Beilstein/REAXYS Number:
3563461
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

97%

impurities

≤3% sodium chloride

mp

120-122 °C (lit.)

SMILES string

SC#N.NC(N)=N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chaotropic agent and strong denaturant; solubilizes cells.

replaced by

Product No.
Description
Pricing

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service