29263
N-(1-Cyclohexenyl)formamide
≥98.0% (GC)
Synonym(s):
1-Formamido-1-cyclohexene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H11NO
CAS Number:
Molecular Weight:
125.17
Beilstein/REAXYS Number:
2828041
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
≥98.0% (GC)
mp
58-63 °C
storage temp.
2-8°C
SMILES string
[H]C(=O)NC1=CCCCC1
Other Notes
Stable, crystalline precursor of the "universal, convertible" 1-cyclohexenyl isocyanide employed in the Ugi four component condensation, 4CC, to give α-(acylamino)amides
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T.A. Keating et al.
Journal of the American Chemical Society, 118, 2574-2574 (1996)
The Journal of Organic Chemistry, 63, 8021-8021 (1998)
C. Hume et al.
Tetrahedron Letters, 39, 1113-1113 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service