About This Item
Recommended Products
vapor pressure
325 mmHg ( 37.7 °C)
assay
≥99%
autoignition temp.
572 °F
refractive index
n20/D 1.392 (lit.)
bp
62 °C (lit.)
mp
−136 °C (lit.)
density
0.682 g/mL at 25 °C (lit.)
SMILES string
CCCC(C)=C
InChI
1S/C6H12/c1-4-5-6(2)3/h2,4-5H2,1,3H3
Inchi Key
WWUVJRULCWHUSA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-16.6 °F - closed cup
flash_point_c
-27 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service