Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

291617

Sigma-Aldrich

HY-STOR® 205 lanthanum-nickel alloy

for hydrogen storage

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:

type

for hydrogen storage

SMILES string

[Ni].[La]

Legal Information

HY-STOR is a registered trademark of Ergenics, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service