Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

289930

Sigma-Aldrich

Mithramycin A from Streptomyces plicatus

90-95%

Synonym(s):

Aureolic acid, Plicamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H76O24
CAS Number:
Molecular Weight:
1085.15
Beilstein/REAXYS Number:
5236667
MDL number:
UNSPSC Code:
12352103

assay

90-95%

SMILES string

[H][C@]1(Cc2cc3cc(O[C@H]4C[C@@H](O[C@H]5C[C@@H](O)[C@H](O)[C@@H](C)O5)[C@@H](O)[C@@H](C)O4)c(C)c(O)c3c(O)c2C(=O)[C@H]1O[C@H]6C[C@@H](O[C@H]7C[C@@H](O[C@H]8C[C@](C)(O)[C@H](O)[C@@H](C)O8)[C@H](O)[C@@H](C)O7)[C@H](O)[C@@H](C)O6)[C@H](OC)C(=O)[C@@H](O)[C@@H](C)O

InChI

1S/C52H76O24/c1-18-29(72-34-14-30(43(58)21(4)68-34)73-33-13-28(54)42(57)20(3)67-33)12-26-10-25-11-27(49(66-9)48(63)41(56)19(2)53)50(47(62)39(25)46(61)38(26)40(18)55)76-36-16-31(44(59)23(6)70-36)74-35-15-32(45(60)22(5)69-35)75-37-17-52(8,65)51(64)24(7)71-37/h10,12,19-24,27-28,30-37,41-45,49-51,53-61,64-65H,11,13-17H2,1-9H3/t19-,20-,21-,22-,23-,24-,27+,28-,30-,31-,32-,33+,34+,35+,36+,37+,41+,42-,43+,44-,45-,49+,50+,51-,52+/m1/s1

InChI key

CFCUWKMKBJTWLW-BGLFSJPPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anticancer antibiotic.
Anticancer antibiotic. Inhibits transcription and protein synthesis. Binds to DNA in native chromatin. Substrate of Pgp in MDR phenotypes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service