289671
4-Nitrophenetole
98%
Synonym(s):
1-Ethoxy-4-nitrobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
O2NC6H4OC2H5
CAS Number:
Molecular Weight:
167.16
Beilstein/REAXYS Number:
972473
EC Number:
MDL number:
UNSPSC Code:
12352103
assay
98%
bp
112-115 °C/3 mmHg (lit.)
mp
54-59 °C (lit.)
SMILES string
CCOc1ccc(cc1)[N+]([O-])=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service