Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

289361

Sigma-Aldrich

Sodium metavanadate

technical grade, 90%

Synonym(s):

Sodium (meta)vanadate, Sodium trioxovanadate, Sodium vanadate(V), Sodium vanadium oxide, Sodium vanadium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaVO3
CAS Number:
Molecular Weight:
121.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

technical grade

assay

90%

reaction suitability

reagent type: catalyst
core: vanadium

SMILES string

[Na+].[O-][V](=O)=O

InChI

1S/Na.3O.V/q+1;;;-1;

InChI key

CMZUMMUJMWNLFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in an ion-exchange process to obtain a porous aerogel of V2O5 with large surface area the aerogel may be intercalated with various alkylmetal reagents.

Replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

target_organs

Respiratory Tract

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le,D.B. et al.
Chemistry of Materials, 10, 682-682 (1998)
Gwendolyn Moise et al.
Biochemistry, 54(42), 6490-6500 (2015-10-08)
Catalysis in protein tyrosine phosphatases (PTPs) involves movement of a protein loop called the WPD loop that brings a conserved aspartic acid into the active site to function as a general acid. Mutation of the tryptophan in the WPD loop
Oluwaseun Mustapha et al.
Environmental toxicology and pharmacology, 38(1), 98-111 (2014-06-14)
Vanadium exposure has been known to lead to lipid peroxidation, demyelination and oligodendrocytes depletion. We investigated behaviour and glial reactions in juvenile mice after early neonatal exposure to vanadium, and examined the direct effects of vanadium in oligodendrocyte progenitor cultures
Izabella Solti et al.
PloS one, 10(6), e0129217-e0129217 (2015-06-09)
Previous studies on the degenerative animal model of multiple sclerosis suggested that the copper-chelator cuprizone might directly suppress T-cell functions. Peripheral T-cell function in the cuprizone model has already been explored; therefore, in the present study, we investigated, for the
Fruzsina Fonai et al.
Biochimica et biophysica acta, 1852(12), 2563-2573 (2015-09-20)
Sepsis caused by LPS is characterized by an intense systemic inflammatory response affecting the lungs, causing acute lung injury (ALI). Dysfunction of mitochondria and the role of reactive oxygen (ROS) and nitrogen species produced by mitochondria have already been proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service